Would you like to log in again?
ReadiUse™ Preactivated HRP Maleimide, AAT Bioquest
Supplier: AAT Bioquest
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
11026
76481-890EA 118.54 USD InStock
76481-890
ReadiUse™ Preactivated HRP Maleimide, AAT Bioquest
Assays Immunological Assays
ReadiUse™ Preactivated maleimide is a thiol-reactive HRP derivative.
  • Ready-to-use
  • Highly useful antibody label
  • Widely used for immunoassays
  • Multiple applications
Caution: For research use only.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty