Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Aminoallyl UTP sodium salt, (AA-UTP) 4 mM in Tris/EDTA (TE) buffer

Supplier: AAT BIOQUEST INC
Warning

17021
76482-816EA 361.89 USD InStock
76482-816
Aminoallyl UTP sodium salt, (AA-UTP) 4 mM in Tris/EDTA (TE) buffer
Aminoallyl UTP sodium salt, (AA-UTP)
The amine-modified uridine 5'-triphosphates (such as aminoallyl-UTP) can be used to produce amine-containing RNA by conventional enzymatic incorporation methods such as transcription, nick translation, random primed labeling, or PCR.


MW: 605.16 g/mol
CAS Number: 75221-88-4
ADR: 5,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty