Would you like to log in again?
Nuclear Orange™ DCS1, AAT Bioquest
Supplier: AAT Bioquest
17551
76482-898EA 118.54 USD InStock
76482-898
Nuclear Orange™ DCS1, AAT Bioquest
Assays Cellular Assays
Our Nuclear Orange™ DCS1 is a fluorogenic, DNA-selective and cell-impermeant dye for analyzing DNA content in dead, fixed or apoptotic cells.
  • Cell-Impermeable
  • Useful For Multicolor Imaging
  • Bright Fluorescence
5 mM DMSO Solution
Caution: For Research Use Only
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty