Would you like to log in again?
FluoroQuest™ Anti-Fading Kit I *Optimized for Slide Imaging* (1 Kit), AAT Bioquest
Supplier: AAT Bioquest
20001
76482-960EA 118.54 USD InStock
76482-960
FluoroQuest™ Anti-Fading Kit I *Optimized for Slide Imaging* (1 Kit), AAT Bioquest
Stains Microscopy and staining reagents
When exposed to excitation light, fluorescence intensity of dyes decreases due to their photooxidation or other photoreactions.
  • Preserves fluorescence
  • Prevents photobleaching
  • Ready to use
  • Contains all necessary components
Caution: For research use only.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty