Would you like to log in again?
Annexin V-mFluor™ Violet 510 Conjugate, AAT Bioquest
Supplier: AAT Bioquest
20081
76483-018EA 240.68 USD InStock
76483-018
Annexin V-mFluor™ Violet 510 Conjugate, AAT Bioquest
Assays Cellular Assays
Annexins are a family of proteins that bind to phospholipid membranes in the presence of calcium.
  • High stability
  • In-house quality control
  • Multiple research applications
  • Optimized for flow cytometry
Caution: For research use only.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty