Would you like to log in again?
Cell Meter™ Live Cell Caspase 1 Binding Assay Kit *Green Fluorescence*, AAT Bioquest
Supplier: AAT Bioquest
20108
76483-028EA 361.89 USD InStock
76483-028
Cell Meter™ Live Cell Caspase 1 Binding Assay Kit *Green Fluorescence*, AAT Bioquest
Assays Cellular Assays
Our Cell Meter™ live cell caspases activity assay kits are based on fluorescent FMK inhibitors of caspases.
  • Based on FMK inhibitor
  • Direct caspase detection
  • Suitable for all major fluorescence platforms
Caution: For research use only.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty