Would you like to log in again?
Cl-NERF (1 mg), AAT Bioquest
Supplier: AAT Bioquest
21220
76483-362EA 240.68 USD InStock
76483-362
Cl-NERF (1 mg), AAT Bioquest
Stains Microbiology Stains
Cl-NERF is a rhodol dye with pKa = 3.8.
  • pH-dependent fluorescence
  • Ratiometric detection
  • Cell impermeable
Caution: For research use only.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty