Would you like to log in again?
Metal Fluor™ Zn-520 potassium salt
Supplier: AAT BIOQUEST INC
21262
76483-380EA 240.68 USD InStock
76483-380
Metal Fluor™ Zn-520 potassium salt
Metal Fluor™ Zn-520 potassium salt
Metal Fluor™ Zn-520 is designed for detection of higher zinc ion concentrations that are present in synaptic vesicles and released in response to electrical stimulation or excitotoxic agonists (0.05 to 50 µM) with minimal interfering calcium sensitivity.
  • Cell impermeable
  • Zinc-dependent fluorescence
  • Multiple applications
Caution: For research use only.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty