Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Thiolite™ Blue, AM (1 mg), AAT Bioquest
Supplier: AAT Bioquest
21506
76483-416EA 118.54 USD InStock
76483-416
Thiolite™ Blue, AM (1 mg), AAT Bioquest
Stains Microbiology Stains
Thiolite™ Blue is one of the most sensitive sensors for measuring thiol compounds.
  • No wash step required
  • Cell permeable
  • Improved sensitivity and signal
  • Optimized for intracellular detection
  • Multiple applications
Caution: For research use only.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty