Would you like to log in again?
MESG *Phosphate Assay Reagent*, AAT Bioquest
Supplier: AAT Bioquest
21600
76483-424EA 212.42 USD InStock
76483-424
MESG *Phosphate Assay Reagent*, AAT Bioquest
Assays Cellular Assays
In the presence of inorganic phosphate MESG is converted to 2-amino-6-mercapto-7-methlpurine by purine nucleoside phosphorylase (EC 2.4.2.1) with absorption wavelength shift to red.
  • Adaptable and robust
  • Quantifiable emission wavelength shift
  • Standalone reagent
Caution: For research use only.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty