Would you like to log in again?
PhosphoWorks™ Luminometric ATP Assay Kit, Steady Glow, AAT Bioquest
Supplier: AAT Bioquest
21608 21609
76483-426EA 1159.15 USD InStock
76483-426 76483-428
PhosphoWorks™ Luminometric ATP Assay Kit, Steady Glow, AAT Bioquest
Assays Cellular Assays
Adenosine triphosphate (ATP) plays a fundamental role in cellular energenics, metabolic regulation and cellular signaling.
  • Extreme sensitivity
  • Stable signal with >4 hour half-life
  • No mixing or separations required
Caution: For research use only.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty