Would you like to log in again?
PhosphoWorks™ Fluorimetric Pyrophosphate Assay Kit, Enhanced Selectivity, AAT Bioquest
Supplier: AAT Bioquest
21614
76483-440EA 423.14 USD InStock
76483-440
PhosphoWorks™ Fluorimetric Pyrophosphate Assay Kit, Enhanced Selectivity, AAT Bioquest
Assays Cellular Assays
Pyrophosphate (PPi) is produced by a number of biochemical reactions, such as ATP hydrolysis, DNA and RNA polymerizations, cyclic AMP formation by the enzyme adenylate cyclase and the enzymatic activation of fatty acids to form their coenzyme A esters.
  • Pyrophosphate-dependent fluorescence
  • Highly selective for PPi
  • Contains all necessary components
Caution: For research use only.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty