Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Calcein UltraBlue™ AM (10×50 µg), AAT Bioquest
Supplier: AAT Bioquest
21908
76483-458EA 240.68 USD InStock
76483-458
Calcein UltraBlue™ AM (10×50 µg), AAT Bioquest
Stains Microbiology Stains
Calcein UltraBlue™ AM is a cell-permeable version of Calcein UltraBlue™.
  • Low cytotoxicity
  • Excellent cell retention
  • Cell permeable
  • Suitable for Multiplexing
Caution: For research use only.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty