Would you like to log in again?
CytoTrace™ Red CFDA, AAT Bioquest
Supplier: AAT Bioquest
22016
76483-486EA 240.68 USD InStock
76483-486
CytoTrace™ Red CFDA, AAT Bioquest
Assays Cellular Assays
CytoTrace™ Red fluorescent probe retains the spectral properties of Cy3/TRITC, thus its signal can be easily monitored using fluorescence microscopy or flow cytometry.
  • High Multigenerational Cell Retention
  • Bright Signal
Caution: For Research Use Only
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty