Would you like to log in again?
C1-Bodi Fluor 500/510-C12 [equivalent to BODIPY® 500/510 C1, C12], AAT Bioquest
Supplier: AAT Bioquest
22120
76483-554EA 240.68 USD InStock
76483-554
C1-Bodi Fluor 500/510-C12 [equivalent to BODIPY® 500/510 C1, C12], AAT Bioquest
Assays Cellular Assays
The green fluorescent BODIPY FL C12 fatty acid can be used as a synthetic precursor to a wide variety of fluorescent phospholipids.
  • Phospholipid Precursor
  • pH-Insensitive Fluorescence
  • Fluorescent In Aqueous and Lipids
Caution: For Research Use Only
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty