Would you like to log in again?
JC-10
Supplier: AAT BIOQUEST INC
Synonyms: Enhanced JC-1
22204
76483-562EA 118.54 USD InStock
76483-562
JC-10
JC-10
JC-10 is capable of entering selectively into mitochondria, and changes reversibly its color from green to orange as membrane potentials increase.
  • Improved water solubility
  • Low precipitation rates
  • Selective for mitochondria
  • Superior performance
When excited at 490 nm, the color of JC-10 changes reversibly from green to greenish orange as the mitochondrial membrane becomes more polarized. Both colors can be detected using the filters commonly mounted in all flow cytometers, so that green emission can be analyzed in fluorescence channel 1 (FL1) and greenish orange emission in channel 2 (FL2).
Caution: For research use only.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty