Would you like to log in again?
Rhodamine B, Hexyl Ester, Perchlorate (10 mg), AAT Bioquest
Supplier: AAT Bioquest
22211
76483-568EA 93.54 USD InStock
76483-568
Rhodamine B, Hexyl Ester, Perchlorate (10 mg), AAT Bioquest
Stains Microbiology Stains
Positively charged rhodamine dyes (such as rhodamine esters and rosamines) are selectively localized in mitochondria, thus they are widely used for labeling mitochondria of live cells.
  • Selective for mitochondria
  • Easily-detectable signal
  • Multiple applications
Caution: For research use only.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty