Would you like to log in again?
CytoTell™ Blue, AAT BIOQUEST INC
Supplier: AAT Bioquest
22252 22251
76483-582EA 180.84 USD InStock
76483-582 76483-580
CytoTell™ Blue, AAT BIOQUEST INC
Assays Cellular Assays
Flow cytometry combined with fluorescence staining is a powerful tool to analyze heterogeneous cell populations.
  • Minimal Cytotoxicity
  • Useful For Multicolor Imaging
Caution: For Research Use Only
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty