Would you like to log in again?
Live or Dead™ Fixable Dead Cell Staining Kit *Red Fluorescence*, AAT Bioquest
Supplier: AAT Bioquest
22603
76483-614EA 180.84 USD InStock
76483-614
Live or Dead™ Fixable Dead Cell Staining Kit *Red Fluorescence*, AAT Bioquest
Assays Cellular Assays
Our Live or Dead™ Fixable Dead Cell Staining Kits are a set of tools for labeling cells for fluorescence microscopic investigations of cell functions.
  • Contains all necessary components
  • Bright and stable signal
  • Suitable for all major fluorescence platforms
Caution: For research use only.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty