Would you like to log in again?
Cell Navigator® Mitochondrion Staining Kit *Orange Fluorescence with 405 nm Excitation* (500 Assays), AAT Bioquest
Supplier: AAT Bioquest
22673
76483-692EA 240.68 USD InStock
76483-692
Cell Navigator® Mitochondrion Staining Kit *Orange Fluorescence with 405 nm Excitation* (500 Assays), AAT Bioquest
Assays Cellular Assays
Our Cell Navigator® fluorescence imaging kits are a set of fluorescence imaging tools for labeling sub-cellular organelles such as membranes, lysosomes, mitochondria, nuclei, etc.
  • Cell-permeant staining
  • Improved signal
  • Suitable for all major fluorescence platforms
Caution: For research use only.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty