Would you like to log in again?
MitoLite™ Blue FX490, AAT Bioquest
Supplier: AAT Bioquest
22674
76483-694EA 118.54 USD InStock
76483-694
MitoLite™ Blue FX490, AAT Bioquest
Assays Cellular Assays
MitoLite™ Blue is a cationic dye that selectively accumulates in mitochondria probably via the mitochondrial membrane potential gradient.
  • Mitochondria-Specific Dye
  • Cell-Permeable
  • Robust
Caution: For Research Use Only
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty