Would you like to log in again?
Live or Dead™ Cell Viability Assay Kit *Green/Red Dual Fluorescence*, AAT Bioquest
Supplier: AAT Bioquest
22760 22789
76483-728EA 970.09 USD InStock
76483-728 76483-730
Live or Dead™ Cell Viability Assay Kit *Green/Red Dual Fluorescence*, AAT Bioquest
Assays Cellular Assays
This Live or Dead™cell viability uses two fluorogenic indicators: calcein AM for viable cells and a cell-impermeable DNA-binding dye for the cells with compromised membranes.
  • Contains all necessary components
  • Multiple research applications
  • Bright and stable signal
  • Suitable for all major fluorescence platforms
  • Results in under 2 hours
Caution: For research use only.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty