Would you like to log in again?
Screen Quest™ Luminometric Calcium Assay Kit, AAT Bioquest
Supplier: AAT Bioquest
36306 36305
76483-924EA 423.14 USD InStock
76483-924 76483-926
Screen Quest™ Luminometric Calcium Assay Kit, AAT Bioquest
Assays Cellular Assays
Calcium flux assays are preferred methods in drug discovery for screening G protein coupled receptors (GPCR).
  • Improved sensitivity
  • Robust and adaptable protocol
  • Multiple research applications
Caution: For research use only.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty