Would you like to log in again?
Screen Quest™ Fluorimetric ELISA cAMP Assay Kit, AAT Bioquest
Supplier: AAT Bioquest
36374 36373
76483-974EA 2376.83 USD InStock
76483-974 76483-972
Screen Quest™ Fluorimetric ELISA cAMP Assay Kit, AAT Bioquest
Assays ELISAs
Screen Quest™ Fluorescence ELISA cAMP Assay Kit provides an optimized assay method for monitoring the activation of adenylyl cyclase in G-protein coupled receptor systems.
  • No acetylation step
  • Single wash step
  • Robust protocol
  • Sensitive substrate for easy quantification
Caution: For research use only.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty