Would you like to log in again?
Screen Quest™ TR-FRET No Wash cAMP Assay Kit, AAT Bioquest
Supplier: AAT Bioquest
36381 36380 36379
76483-976EA 604.2 USD InStock
76483-976 76483-978 76483-980
Screen Quest™ TR-FRET No Wash cAMP Assay Kit, AAT Bioquest
Assays Cellular Assays
Screen Quest™ TR-FRET No Wash cAMP Assay Kit provides a convenient assay method for monitoring the activation of adenylyl cyclase in G-protein coupled receptor systems.
  • Extremely sensitive detection
  • Contains all necessary components
  • No wash step required
  • Robust and adaptable protocol
  • Longer fluorescence lifetime
Caution: For research use only.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty