Would you like to log in again?
Screen Quest™ Live Cell cAMP Assay Service Pack, AAT Bioquest
Supplier: AAT Bioquest
36384 36383 36382
76483-986EA 5571.59 USD InStock
76483-986 76483-984 76483-982
Screen Quest™ Live Cell cAMP Assay Service Pack, AAT Bioquest
Assays Cellular Assays
G protein coupled receptors (GPCR) are one of the largest receptor classes targeted by drug discovery programs.
  • Extremely sensitive detection
  • Contains all necessary components
  • No lysis step required
  • Robust and adaptable protocol
  • Longer fluorescence lifetime
Caution: For research use only.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty