Would you like to log in again?
Cell Meter™ FITC-Annexin V Binding Apoptosis Assay Kit *Optimized for Flow Cytometry*, AAT Bioquest
Supplier: AAT Bioquest
22839
76484-130EA 240.68 USD InStock
76484-130
Cell Meter™ FITC-Annexin V Binding Apoptosis Assay Kit *Optimized for Flow Cytometry*, AAT Bioquest
Assays Cellular Assays
Our Cell Meter™ FITC-Annexin V binding assay kit is designed to monitor cell apoptosis through measuring the translocation of phosphatidylserine (PS).
  • Robust protocol
  • Sensitive detection
  • In-house quality control
Caution: For research use only.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty