Would you like to log in again?
Live or Dead™ Cell Viability Assay Kit *Red/Blue Dual Fluorescence*, AAT Bioquest
Supplier: AAT Bioquest
22788
76484-404EA 361.89 USD InStock
76484-404
Live or Dead™ Cell Viability Assay Kit *Red/Blue Dual Fluorescence*, AAT Bioquest
Assays Cellular Assays
This Live or Dead™ Cell Viability Assay Kit uses two fluorescent indicators: Cellbrite™ Red (Ex/Em = 613/631 nm) for labeling viable cells and a cell-impermeable DNA-binding dye Nuclear Blue™ DCS1 (Ex/Em = 360/450 nm) for labeling dead cells with damaged membranes.
  • Contains all necessary components
  • Multiple research applications
  • Bright and stable signal
  • Suitable for all major fluorescence platforms
  • Results in under 2 hours
Caution: For research use only.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty