Would you like to log in again?
Screen Quest™ Calbryte-520 Probenecid-Free and Wash-Free Calcium Assay Kit, AAT Bioquest
Supplier: AAT Bioquest
36319 36317 36318
76484-406EA 240.68 USD InStock
76484-406 76484-408 76484-410
Screen Quest™ Calbryte-520 Probenecid-Free and Wash-Free Calcium Assay Kit, AAT Bioquest
Assays Cellular Assays
Calcium flux assays are the preferred methods in drug discovery for screening G protein coupled receptors (GPCR).
  • 100 to 250 times fluorescence increase once bound to calcium
  • No probenecid required
  • Contains all necessary components
  • No wash step required
  • Robust and adaptable protocol
  • Brighter and more photostable signal
Caution: For research use only.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty