Would you like to log in again?
Buccutite™ Rapid APC-iFluor® 700 Tandem Antibody Labeling Kits (2 Labelings), AAT Bioquest
Supplier: AAT Bioquest
1319 1347
76484-456EA 361.89 USD InStock
76484-456 76484-442
Buccutite™ Rapid APC-iFluor® 700 Tandem Antibody Labeling Kits (2 Labelings), AAT Bioquest
Protein Linking and Labeling Systems
APC-iFluor® 700 Tandem is an excellent replacement for APC-Alexa Fluor® 700 Tandem since they have almost identical spectra.
  • High conjugation yield
  • Stable crosslinking
  • Minimal hands-on time
  • Robust protocol
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty