Would you like to log in again?
Cell Navigator® Fluorimetric Lipid Droplet Assay Kit *Red Fluorescence*, AAT Bioquest
Supplier: AAT Bioquest
22735
76484-486EA 240.68 USD InStock
76484-486
Cell Navigator® Fluorimetric Lipid Droplet Assay Kit *Red Fluorescence*, AAT Bioquest
Assays Cellular Assays
Lipid droplets, also referred to as lipid bodies, oil bodies or adiposomes, are lipid-rich cellular organelles that regulate the storage and hydrolysis of neutral lipids.
  • Live cell labeling
  • Improved signal
  • Suitable for all major fluorescence platforms
Caution: For research use only.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty