Would you like to log in again?
MycoLight™ Ratiometric Bacterial Membrane Potential Kit *Red/Green Fluorescence*, AAT Bioquest
Supplier: AAT Bioquest
22401
76484-498EA 361.89 USD InStock
76484-498
MycoLight™ Ratiometric Bacterial Membrane Potential Kit *Red/Green Fluorescence*, AAT Bioquest
Assays Immunological Assays
AAT Bioquest's MycoLight™ Ratiometric Bacterial Membrane Potential Kit uses a fluorescent sensor that exhibits green fluorescence in both gram-positive and negative bacterial cells in low concentration, but the fluorescence shifts toward red emission at higher cytosolic concentrations due to the dye molecule aggregation caused by larger membrane potentials.
  • Suitable for gram-positive and gram-negative bacteria
  • Ratiometric detection
  • Contains all necessary Components
  • Multiple applications
Caution: For research use only.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty