Would you like to log in again?
Portelite™ Fluorimetric Protein Quantitation Kit, Optimized for CytoCite™ and Qubit™ Fluorometers, AAT Bioquest
Supplier: AAT Bioquest
11111
76484-588EA 240.68 USD InStock
76484-588
Portelite™ Fluorimetric Protein Quantitation Kit, Optimized for CytoCite™ and Qubit™ Fluorometers, AAT Bioquest
Assays Protein Assays
Protein quantification is an essential task in protein purification, electrophoresis, cell biology, molecular biology and other research applications.
  • Dynamic range from 12.5 ug/ml to 5 mg/ml of bsa
  • Optimized for cytocite™ and qubit™ fluorometers
  • Highly sensitive
  • Robust protocol
Caution: For research use only.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty