Would you like to log in again?
Cell Meter™ Intracellular Colorimetric Lipid Peroxidation (MDA) Assay Kit, AAT Bioquest
Supplier: AAT Bioquest
15991
76484-672EA 483.08 USD InStock
76484-672
Cell Meter™ Intracellular Colorimetric Lipid Peroxidation (MDA) Assay Kit, AAT Bioquest
Assays Cellular Assays
Lipid peroxidation is characterized by the oxidative degradation of unsaturated fatty acids, phospholipids, glycolipids, cholesterol esters and cholesterol.
  • MDA-specific
  • Robust protocol
  • No high temperatures required
Caution: For research use only.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty