Would you like to log in again?
MycoLight™ Flow Cytometric Live Bacteria Assay Kit, AAT Bioquest
Supplier: AAT Bioquest
22407
76484-740EA 361.89 USD InStock
76484-740
MycoLight™ Flow Cytometric Live Bacteria Assay Kit, AAT Bioquest
Assays Immunological Assays
The MycoLight Flow cytometric live bacteria assay kit provides an easy and convenient method for evaluating bacterial vitality as a function of the intracellular esterase activity.
  • Suitable for gram-positive and gram-negative bacteria
  • Contains all necessary components
  • Multiple applications
  • Optimized for flow cytometry
Caution: For research use only.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty