Would you like to log in again?
MitoLite™ Red CMXRos, AAT Bioquest
Supplier: AAT Bioquest
22698
76484-840EA 180.84 USD InStock
76484-840
MitoLite™ Red CMXRos, AAT Bioquest
Assays Cellular Assays
MitoLite™ Red CMXRos is chemically same to the MitoTracker™ Red CMXRos (ThermoFisher).
  • Mitochondria-Specific Dye
  • Cell-Permeable
  • Robust
Caution: For Research Use Only
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty