Would you like to log in again?
PSA™ iFluor® 555 Imaging Kit with Goat Anti-Mouse IgG
Supplier: AAT Bioquest
PSA™
45270
76484-928EA 604.2 USD InStock
76484-928
PSA™ iFluor® 555 Imaging Kit with Goat Anti-Mouse IgG
Protein Linking and Labeling Systems
iFluor® 555 PSA kit is a much superior replacement for Alexa Fluor 555 tyramide-based kit or other spectrally similar fluorescent tyramide or TSA kits.
  • Higher efficiency
  • Improved fluorescence signal
  • Less consumption of primary antibody
  • 10 to 50X improvement over TSA reagents
In combination with our superior iFluor® dyes that have higher florescence intensity, increased photostability and enhanced water solubility, the iFluor® dye-labeled Styramide™ conjugates can generate fluorescence signal with significantly higher precision and sensitivity (more than 100 times) than standard ICC/IF/IHC.
Packaging: Plastic vial.
Caution: For research use only.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty