Would you like to log in again?
N-Me-DMAE NHS ester
Supplier: AAT BIOQUEST INC
26005
76484-992EA 240.68 USD InStock
76484-992
N-Me-DMAE NHS ester
N-Me-DMAE NHS ester
The acridinium NHS ester can be used to label proteins and nucleic acids.
  • Chemiluminescent labeling
  • Excellent sensitivity
  • Suitable for various Experimental surfaces
  • Multiple applications
Caution: For research use only.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty