Would you like to log in again?
Calcein Deep Red™ AM ester
Supplier: AAT BIOQUEST INC
22011
76485-142EA 240.68 USD InStock
76485-142
Calcein Deep Red™ AM ester
Calcein Deep Red™ AM ester
Calcein AM is one the most popular fluorescent probes used for labeling and monitoring cellular functions of live cells.
  • Low cytotoxicity
  • Excellent cell retention
  • Cell permeable
  • Suitable for multiplexing
Caution: For research use only.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty