Would you like to log in again?
CytoFix™ Red Mitochondrial Stain, AAT Bioquest
Supplier: AAT Bioquest
23200
76485-166EA 361.89 USD InStock
76485-166
CytoFix™ Red Mitochondrial Stain, AAT Bioquest
Assays Cellular Assays
CytoFix™ Red mitochondrial stain is a dye that selectively stains mitochondria independent of mitochondrial membrane potential gradient.
  • Mitochondria-Specific Dye
  • Cell-Permeable
  • Useful for Multicolor Imaging
  • Robust
  • Retained After Fixation
Caution: For Research Use Only
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty