Would you like to log in again?
TRITC-Dextran Conjugate (Average MW = ~70K), AAT Bioquest
Supplier: AAT Bioquest
21708
76485-180EA 229.01 USD InStock
76485-180
TRITC-Dextran Conjugate (Average MW = ~70K), AAT Bioquest
Assays Cellular Assays
TRITC-labeled dextran is used in cardiovascular, microcirculation, perfusion, cell monolayer and cell membrane permeability research as fluorescent flux tracer compound that supports the measurement of processes such as blood flow, membrane damage, vascular drainage and renal elimination.
  • High quality
  • Multiple applications
Caution: For research use only.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty