Would you like to log in again?
Cell Meter™ BX520 Fixable Viability Dye, AAT Bioquest
Supplier: AAT Bioquest
22510
76485-252EA 229.01 USD InStock
76485-252
Cell Meter™ BX520 Fixable Viability Dye, AAT Bioquest
Assays Cellular Assays
Discrimination and exclusion of dead cells from live cells allows cleaner separation and identification of cell populations.
  • Cell-Impermeable
  • Amine- And Thiol-Reactive
  • High Stability
  • Live/Dead Cell Labeling Dye
  • Fixable
Caution: For Research Use Only
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty