Would you like to log in again?
ReadiUse™ dNTP Mix *10 mM* (5 ml), AAT Bioquest
Supplier: AAT Bioquest
ReadiUse™
17200
76485-404EA 344.92 USD InStock
76485-404
ReadiUse™ dNTP Mix *10 mM* (5 ml), AAT Bioquest
Nucleic Acid Reagents Nucleotides
Our Ready-to Use Solution of deoxynucleotide triphosphates (dNTPs) have been extensively tested and verified for use in a wide variety of molecular biology applications, including PCR, real-time PCR, high-fidelity PCR, long PCR, cDNA synthesis, RT-PCR, RCA, MDA, DNA labeling, DNA sequencing, in vitro transcription, siRNA synthesis, and RNA amplification.
  • Ready to use formulation
  • In-house testing and quality control
  • Multiple applications
Caution: For research use only.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty