Would you like to log in again?
ReadiLink™ xtra Rapid Cy3 Antibody Labeling Kit *BSA-Compatible* (2 Labelings), AAT Bioquest
Supplier: AAT Bioquest
1971
76485-434EA 229.01 USD InStock
76485-434
ReadiLink™ xtra Rapid Cy3 Antibody Labeling Kit *BSA-Compatible* (2 Labelings), AAT Bioquest
Protein Linking and Labeling Systems
ReadiLink™ xtra rapid antibody labeling kits require essentially only 2 simple mixing steps without a column purification needed.
  • Contains all essential components
  • Minimal hands on time
  • Ready to use
  • Robust and simple antibody labeling protocol
  • No column purification required
Caution: For research use only.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty