Would you like to log in again?
Tetracysteine FLASH Diacetate *2 mM DMSO Solution*, AAT Bioquest
Supplier: AAT Bioquest
22334
76485-504EA 447.08 USD InStock
76485-504
Tetracysteine FLASH Diacetate *2 mM DMSO Solution*, AAT Bioquest
Assays Protein Assays
FLASH diacetate is a cell-permeable version of FlASH, which is a fluorescein derivative, modified to contain two arsenic atoms at a set distance from each other.
  • Fluorescein derivative
  • High affinity for thiols
  • Multiple applications
Caution: For research use only.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty