Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cell Meter™ Cell Adhesion Assay Kit, AAT Bioquest

Supplier: AAT Bioquest
23010
76485-524EA 402.7 USD InStock
76485-524
Cell Meter™ Cell Adhesion Assay Kit, AAT Bioquest
Assays Cellular Assays
The Cell Meter™ Cell Adhesion Assay Kit is a fast and sensitive assay for measuring cell-cell or cell-surface adhesion for a variety of cell types.

  • Contains all necessary components
  • Suitable for most common cell types
  • pH-independent fluorescence
  • Multiple research applications
  • Bright and stable signal
  • Suitable for all major fluorescence platforms

Caution: For research use only.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty