Would you like to log in again?
Cell Meter™ Fluorescence Gap Junction Tracing Kit, AAT Bioquest
Supplier: AAT Bioquest
23600
76485-538EA 459.42 USD InStock
76485-538
Cell Meter™ Fluorescence Gap Junction Tracing Kit, AAT Bioquest
Assays Cellular Assays
Gap junctions are specialized intercellular membrane channels constituted with connexin that selectively facilitate the passage of small molecules of <1.5 Kd across cells.
  • Contains all necessary components
  • Multiple research applications
  • Bright and stable signal
  • Suitable for all major fluorescence platforms
Caution: For research use only.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty