Would you like to log in again?
HRP-Biotin Conjugate, AAT Bioquest
Supplier: AAT Bioquest
11022
76485-666EA 111.91 USD InStock
76485-666
HRP-Biotin Conjugate, AAT Bioquest
Assays Immunological Assays
Biotinylated horse radish peroxidase (HRP) is an excellent tool for biological detections with ELISA and other enzyme-coupled assays.
  • Highly useful antibody label
  • Widely used for immunoassays
  • Multiple applications
Caution: For research use only.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty