Would you like to log in again?
CytoWatch™ QZ100 Monocyte Blocking Reagent, AAT Bioquest
Supplier: AAT Bioquest
37001 37000
76485-718EA 446.71 USD InStock
76485-718 76485-716
CytoWatch™ QZ100 Monocyte Blocking Reagent, AAT Bioquest
Assays Cellular Assays
Some dye-labeled fluorescent antibody conjugates used in cell surface staining of live cells often exhibit non-specific binding in monocytes and macrophages.
  • Suitable for PBMC or whole blood
  • Minimizes background signal
  • No effect on lymphocytes, monocytes, or granulocytes
Caution: For research use only.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty