Would you like to log in again?
Stainless Steel Gowning Benches, Metro
Supplier: Metro International
GB1636S GB948S GB1236S GB1672S GB1660S GB1672S GB1248S GB1648S GB1660S GB1636S GB960S GB936S GB1224S GB1648S GB1272S GB972S GB1260S
76485-948EA 1652.53 USD InStock
76485-948 76485-946 76485-958 CA19766-782 CA19766-780 19766-782 19766-780 76485-944 76485-956 76485-954 CA19766-778 19766-778 CA19766-776 76485-952 19766-776 76485-960 76485-950
Stainless Steel Gowning Benches, Metro
Furniture Gowning Benches
Type 304 stainless steel, H.D. Super® Posts and proven Super Erecta construction throughout.
  • Heavy-duty, solid top bench
  • Sturdy and comfortable
  • Type 304 stainless steel
  • Adjustable stainless steel foot plates allow floor mounting
  • Non-conductive, but can be made conductive with the addition of HD aluminum split sleeves
Heavy-duty, solid top bench provides a sturdy and comfortable growning fixture. Top is Type 304 stainless steel. Adjustable stainless steel foot plates allow floor mounting. Floor mounting required for 9 and 12" wide benches. All benches are 18" (457mm) tall. Unit is non-conductive, but can be made conductive with the addition of HD aluminum split sleeves.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty